Learn More
BIG SCIENCE INC GIANT PLAIN STIR BAR 6IN 1IN D
Catalog No. NC1969479
Encompass
Giant Plain Stir bar: Length 159 mm (6 inches), Diameter 27 mm (1 inch)
Catalog No. NC1969479 Supplier BIG SCIENCE INC Supplier No. XLSBM15927PLA
Giant Plain Stir bar: Length 159 mm (6 inches), Diameter 27 mm (1 inch)