Learn More
Chemglass Life Sciences Stir Bar, Giant, Magnetic, PTFE, Length x Diameter: 108 x 27mm / 4-1/4 x 1-1/16 inches
SDP
Catalog No. CG201021
Encompass_Preferred
Stir Bar, Giant, Magnetic, PTFE, Length x Diameter: 108 x 27mm / 4-1/4 x 1-1/16 inches. This heavy duty PTFE stir bar is capable of mixing volumes up to a 50 gallon drum. Perfect for use with carboys up to 12 gallon. The bar will fit into a minimum neck diameter of 32mm. Manufactured with FDA-grade and USP Class VI PTFE.
Catalog No. CG201021 Supplier Chemglass Life Sciences Supplier No. CG201021
Stir Bar, Giant, Magnetic, PTFE, Length x Diameter: 108 x 27mm / 4-1/4 x 1-1/16 inches. This heavy duty PTFE stir bar is capable of mixing volumes up to a 50 gallon drum. Perfect for use with carboys up to 12 gallon. The bar will fit into a minimum neck diameter of 32mm. Manufactured with FDA-grade and USP Class VI PTFE.