Learn More
AdipoGen anti-Giantin, mAb (rec.) (TA10) (ATTO 488) (100 µg)
SDP
Supplier:  AdipoGen AG27B0003TDC100
Encompass_Preferred
Recombinant Antibody. Recognizes human and mouse giantin. Species cross-reactivity: Human, Mouse. Clone: TA10. Isotype: Human IgG2lambda. Applications: ICC. Host: Purified from HEK 293 cell culture supernatant. Liquid. In PBS containing 0.02% sodium azide. Giantin, the largest Golgi membrane protein, is C-terminally anchored to cis- and medial-Golgi membranes and forms a long ~250nm coiled-coil rod that directly interacts with microtubules in a cell-cycle regulated way. It is an excellent marker for mammalian golgi complex. It is also an antigen in chronic rheumatoid arthritis and in the autoimmune disease Sjoegren syndrome.
Catalog No. 50-205-3019