Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECHLAB™ GIARDIA/CRYPTOSPORIDIUM CHEK

Catalog No. 23900560
Change view
Click to view available options
Detectable Analytes:
Giardia Cyst Antigen, Cryptosporidium Oocyst Antigen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes
23-900-560 Giardia Cyst Antigen, Cryptosporidium Oocyst Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-560 Supplier TECHLAB™ Supplier No. 30401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Enzyme immunoassay for the qualitative detection of Giardia cyst antigen and Cryptosporidium oocyst antigen in human specimens.

Enzyme immunoassay for the qualitative detection of Giardia cyst antigen and Cryptosporidium oocyst antigen in human fecal specimens as an aid in the diagnosis of patients with diarrhea suspected of Giardia and/or Cryptosporidium gastrointestinal infections.

  • Results in <2 hrs
  • High specificity and sensitivity

Specifications

CPT Code 87449
CE Marker CE-IVD
Clia Complexity High
Detectable Analytes Giardia Cyst Antigen, Cryptosporidium Oocyst Antigen
Detection Method Colorimetric
Format 96-well Plate
Sample Type Stool
Sensitivity 0.976
Disposable Single-use
For Use With (Application) ELISA
Specificity 1
Sterility Non-sterile
Test Time <2 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.