Learn More
ATCC Giardia intestinalis; Portland-1; genomic DNA
Supplier:  ATCC 30888D
Encompass
Genomic DNA isolated from Giardia intestinalis Portland-1 (ATCC 30888). This product can be used in PCR and other molecular biology applications.
Catalog No. 50-238-3576
 Restricted Item