Learn More
ATCC Giardia intestinalis; WB
Catalog No. 502384093
Encompass
Giardia intestinalis strain WB is a parasitic protozoan that was isolated from duodenal aspirate in Bethesda, Maryland. This strain has applications in agricultural research, enteric disease research, and infectious disease research.
Catalog No. 50-238-4093 Supplier ATCC Supplier No. 30957
Giardia intestinalis strain WB is a parasitic protozoan that was isolated from duodenal aspirate in Bethesda, Maryland. This strain has applications in agricultural research, enteric disease research, and infectious disease research.