Learn More
Cayman Chemical GIbberelIn A4 10mg
SDP
Catalog No. 502990677
Encompass_Preferred
A plant hormone with plant signaling and growth stimulatory activities; found in the micropylar endothelium cells of Arabidopsis maternal seed tissue and triggers suspensor PCD in the embryo; increases hypocotyl growth of cucumber seedlings at 6-100 µM; increases flowering in apple trees when applied post-flowering in the previous year
Catalog No. 50-299-0677 Supplier Cayman Chemical Supplier No. 3140010 MG
A plant hormone with plant signaling and growth stimulatory activities; found in the micropylar endothelium cells of Arabidopsis maternal seed tissue and triggers suspensor PCD in the embryo; increases hypocotyl growth of cucumber seedlings at 6-100 µM; increases flowering in apple trees when applied post-flowering in the previous year