Learn More
Gold Biotechnology Inc Gibberellic Acid 3 (Quick-Dissolve) 10 g
SDP
Catalog No. NC2943346
Encompass_Preferred
Gibberellic Acid, also called Gibberellin A3, is a plant hormone in the family of gibberellins, a large class of naturally ocuring tetracyclic diterpene acids which promote cell growth and development. In small concentrations, GA3 is a potent, dihydroxylated gibberellin which commonly used in the triggering of seed germination, stem differentiation and plant maturation.GA3 is widely used to promote the production of larger grape bunches or the induction of other fruit without pollination or seed development. Gibberellic Acid 3 is a growth regulator.
Catalog No. NC2943346 Supplier Gold Biotechnology Inc Supplier No. G12010
Gibberellic Acid, also called Gibberellin A3, is a plant hormone in the family of gibberellins, a large class of naturally ocuring tetracyclic diterpene acids which promote cell growth and development. In small concentrations, GA3 is a potent, dihydroxylated gibberellin which commonly used in the triggering of seed germination, stem differentiation and plant maturation.GA3 is widely used to promote the production of larger grape bunches or the induction of other fruit without pollination or seed development. Gibberellic Acid 3 is a growth regulator.