Learn More
Indofine Chemical Gibberellic Acid, 90%, 5 Gm
SDP
Catalog No. 50908892
Encompass_Preferred
GIBBERELLIC ACID 5 GM, Rare Organics & Biochemicals, Gibberellin A3, C19H22O6, MW: 346.4, >90% by HPLC, 77-06-5, MFCD00079329
Catalog No. 50-908-892 Supplier Indofine Chemical Supplier No. BIO129 5 GM
GIBBERELLIC ACID 5 GM, Rare Organics & Biochemicals, Gibberellin A3, C19H22O6, MW: 346.4, >90% by HPLC, 77-06-5, MFCD00079329