Learn More
Medchemexpress LLC Gibberellic acid | 77-06-5 | 346.37 | 1 ML
SDP
Catalog No. 5000416181
Encompass_Preferred
Gibberellic Acid, named after the fungus *Gibberella fujikuroi*, plays a crucial role in regulating various processes in plant development and growth. It enhances plant response to environmental stresses such as cold, drought, and heavy metals. Additionally, it causes increased lipid peroxidation and fluctuations in the antioxidant defense system in rats.
  • Regulates plant development and growth.
  • Enhances stress response to environmental factors.
  • Supports seed development and germination.
  • Promotes stem and root growth.
  • Involved in cell division and flowering time.
Catalog No. 50-004-16181 Supplier Medchemexpress LLC Supplier No. HYN196410MM/1ML
Gibberellic Acid, named after the fungus *Gibberella fujikuroi*, plays a crucial role in regulating various processes in plant development and growth. It enhances plant response to environmental stresses such as cold, drought, and heavy metals. Additionally, it causes increased lipid peroxidation and fluctuations in the antioxidant defense system in rats.
  • Regulates plant development and growth.
  • Enhances stress response to environmental factors.
  • Supports seed development and germination.
  • Promotes stem and root growth.
  • Involved in cell division and flowering time.