Learn More
Alkali Scientific Gibberellic Acid A3 [GA3], 1 G
SDP
Catalog No. 5000659950
Encompass_Preferred
Gibberellic Acid A3 (GA3) is a plant growth regulator available in powder form, with a 1g package size. GA3 is used to promote cell elongation and germination in plant tissues, helping to overcome dormancy in seeds and stimulate growth in various plant species. It is widely used in plant tissue culture to enhance the growth of shoots, roots, and other plant structures. In addition to its applications in research, GA3 is used in the agricultural industry to increase crop yields and improve fruit development. The powder form allows for convenient storage and accurate preparation of solutions for laboratory and commercial use.
Catalog No. 50-006-59950 Supplier Alkali Scientific Supplier No. GIB001CP
Gibberellic Acid A3 (GA3) is a plant growth regulator available in powder form, with a 1g package size. GA3 is used to promote cell elongation and germination in plant tissues, helping to overcome dormancy in seeds and stimulate growth in various plant species. It is widely used in plant tissue culture to enhance the growth of shoots, roots, and other plant structures. In addition to its applications in research, GA3 is used in the agricultural industry to increase crop yields and improve fruit development. The powder form allows for convenient storage and accurate preparation of solutions for laboratory and commercial use.