Learn More
Bioworld Gibberellic Acid A3, 5 g
Supplier:  Bioworld 306310082
Encompass_Preferred
Gibberellic acid A3 (GA3)A3 content: >93.0% of total gibberellinsPromotes shoot elongation, breaks certain dormancies in seedsPlant cell culture testedAppearance: white to off-white powderMelting Point: 227 °CSoluble in ethanol
Catalog No. 50-255-043