Learn More
Bioworld Gibberellic Acid A3 (HPLC), 1 g
Supplier:  Bioworld 306310091
Encompass_Preferred
Gibberellic acid (HPLC)Synonyms: Gibberellin A3, GA3Promotes shoot (internode) elongation; Breaks certain dormancies in seeds90% gibberellin A3 basis (HPLC)Plant cell culture testedAppearance: white to yellow-white solidMelting Point: 227 °CSoluble in ethanolHPLC - increased purity >98%
Catalog No. 50-255-044
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.