Learn More
Medchemexpress LLC Gibberellin A1 | 545-97-1 | 1 MG
SDP
Catalog No. 5000418734
Encompass_Preferred
Gibberellin A1 is a type of plant hormone, specifically a C19-gibberellin, isolated from the immature seeds of Phaseolus multiflorus. It acts as a growth-promoting acid and is recognized as the main endogenous hormone responsible for controlling stem elongation in plants. Research indicates its ability to reverse stem growth inhibition.
  • Plant hormone
  • Growth-promoting acid
  • Controls stem elongation
  • Isolated from immature seeds of Phaseolus multiflorus
Catalog No. 50-004-18734 Supplier Medchemexpress LLC Supplier No. HYN74431MG
Gibberellin A1 is a type of plant hormone, specifically a C19-gibberellin, isolated from the immature seeds of Phaseolus multiflorus. It acts as a growth-promoting acid and is recognized as the main endogenous hormone responsible for controlling stem elongation in plants. Research indicates its ability to reverse stem growth inhibition.
  • Plant hormone
  • Growth-promoting acid
  • Controls stem elongation
  • Isolated from immature seeds of Phaseolus multiflorus