Learn More
Selleck Chemical LLC Gibberellins A4/A7-25mg
SDP
Supplier:  Selleck Chemical LLC S933325MG
Encompass_Preferred
Gibberellins A4/A7 (Gibberellin A4 and A7) is the Gibberellin A4 Mixture With Gibberellin A7. Gibberellins A4/A7 promotes internode elongation & breaks certain dormancies in seeds. Gibberellins A4/A7 is an endogenous plant growth regulator, promotes the growth and elongation of cells.
Catalog No. 50-238-9750