Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ AmnioMAX™ C-100 Supplement
GREENER_CHOICE

Catalog No. 12556023
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 mL
75 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-556-023 75 mL
12-556-015 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-556-023 Supplier Gibco™ Supplier No. 12556023
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

A part of kit used to prepare fully supplemented Gibco AmnioMAX C-100 Complete Medium developed for short term culture of human amniotic fluid cells for cytogenetic studies and in vitro diagnostic procedures

Easy-to-use format: Gibco AmnioMAX C-100 Complete Medium is a two-part kit for preparation of a complete medium that requires no additional supplementation. The complete medium can be stored at 2-8°C for up to ten days

  • Quality and performance testing
  • Every lot of Gibco AmnioMAX C-100 Complete Medium is performance tested by a certified US reference cytogenetics laboratory to ensure consistently superior performance
  • Pooled primary human amniotic fluid samples are cultured for six days in Gibco AmnioMAX C-100 Complete Medium before measuring the total number of colonies and total number of mitotic colonies
  • In addition, each lot is tested for sterility, pH and osmolality
  • Unique, optimized formulation
  • Gibco AmnioMAX C-100 Complete Medium contains Fetal Bovine Serum (FBS), gentamicin, and L-glutamine to maximize cell attachment and growth
  • It has enhanced buffering system that provides greater pH stability during culture manipulations

Amniotic Fluid and Chorionic Villus Culture, Cell Analysis, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Clinical, Clinical and Translational Research, Cytogenetics, Mammalian Cell Culture

Order Info

Shipping Condition: Dry Ice

Compliance

Gibco AmnioMAX C-100 Supplement is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: -5 to -20°C
Shipping conditions: Frozen
Shelf life: 16 months from date of manufacture. Use complete medium within 7–10 days once opened.
Form Frozen
Product Type C-100 Supplement
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine
Product Line AmnioMAX
Quantity 75 mL
Shipping Condition Dry Ice
WARNING: Cancer - www.P65Warnings.ca.gov

In vitro diagnostic

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.