Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ B-27™ Plus Supplement (50X)

Catalog No. a3582801
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3582801 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3582801 Supplier Gibco™ Supplier No. A3582801
Only null left
Add to Cart
Edge
Add to Cart

B-27 Plus Supplement is the next-generation of the most cited neuronal cell culture product, B-27 Supplement

B-27 Plus Supplement is a serum-free supplement that provides the highest rate of in vitro survival of primary rodent and human stem cell-derived neurons when used with Neurobasal Plus Medium. B-27 Plus Supplement features an optimized formulation, upgraded manufacturing process, and more stringent quality control of raw materials and final product. These improvements enable the highest rate of neuronal cell survival, improved electrophysiological activity, and enhanced functional maturity versus other neuronal cell culture media systems.


B-27 Plus Supplement, when combined with Neurobasal Plus Medium, offers:

  • At least 50% higher survival rate of primary rat neurons compared to classic B-27 Supplement and Neurobasal Medium
  • Highest survival rate of primary rat, mouse, and human stem cell-derived neurons compared to other commercially available serum-free neuronal media systems
  •  Improved electrophysiological activity and maturation—more consistent, stable, and synchronized spontaneous activity in primary rodent neurons after 10 days compared to BrainPhys™ Neuronal Medium
  •  Accelerates neurite outgrowth and increases the number of synaptic-positive puncta

Available as a 50X, 10 mL supplement, B-27 Plus Supplement can seamlessly replace other neuronal cell culture supplements, such as classic B-27 Supplement. The B-27 Plus Neuronal Culture System is also available for purchase, which is comprised of B-27 Plus Supplement and Neurobasal Plus Medium, offering a convenient and valuable all-in-one ordering solution.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5 to -20°C. Protect from light.
Cell Type Neural Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Tested For Endotoxin, Performance, Sterility
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.