Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Bottle, 100 mL
GREENER_CHOICE

Catalog No. 10339001
Encompass_Preferred
Change view
Click to view available options
Quantity:
264 bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-339-001 264 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-339-001 Supplier Gibco™ Supplier No. 10339001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Gibco™ bottle was designed to provide better ergonomic features, ease of use, and reduced chance of contamination.

This Gibco™ bottle was designed to provide better ergonomic features, ease of use, and reduced chance of contamination. The compact design of the Gibco™ media bottle allows for easier handling and storage as well as reduced packaging. This bottle is sterile, empty, and ready for use in cell culture applications.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 100 mL
Material PET
Quantity 264 bottles
Sterility Sterile
Product Line Gibco
Content And Storage Sterile 264 x 100 mL bottles irradiated to 10-6 sterility.

Store at room temperature.
Shipping Condition Room Temperature
Mouth Wide
Product Type Media Bottle

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.