Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CF1 Mouse Embryonic Fibroblasts, irradiated

Catalog No. A34180
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 x 106 cells
4 x 106 cells
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A34180 2 x 106 cells
A34181 4 x 106 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A34180 Supplier Gibco™ Supplier No. A34180
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Irradiated Gibco CF1 Mouse Embryonic Fibroblasts (MEFs) are feeder cells that are ideal for supporting healthy undifferentiated human and mouse embryonic stem cells (ESCs) and induced pluripotent stem cells (iPSCs) in culture. Manufactured by MTI-GlobalSt

  • Help save precious time with pre-inactivated, ready-to-use MEFs
  • Culture ESCs and iPSCs with confidence using meticulously tested feeder cells
  • Rely on a widely-used product
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Cryopreserved mouse embryonic fibroblasts

Store in liquid nitrogen.
Age Embryonic
Cell Type Feeder Cells (Mouse Embryonic Fibroblast)
Form Cryopreserved
Species Mouse
Donor Source Pooled
Quantity 2 x 106 cells
Shipping Condition Shipped on dry ice
What is the difference between Mouse (ICR) Inactivated Embryonic Fibroblasts (Cat. No. A24903) and CF1 Mouse Embryonic Fibroblasts, irradiated (Cat. Nos. A34180, A34181)? Can they be used to reprogram fibroblasts?

Mouse (ICR) Inactivated Embryonic Fibroblasts and CF1 Mouse Embryonic Fibroblasts, irradiated are inactivated feeder layer fibroblast cells that are isolated from different mouse strains. Researchers often have a preference for one or the other. Neither of these can be used to reprogram fibroblasts.

Is it better to use irradiated mouse embryonic fibroblasts (MEFs) or Mitomycin C-treated MEFs to grow embryonic stem cells?

Either method will work in arresting cell division. However, the irradiation process will ensure that cell division will cease regardless of cell aggregation. Cell clumping can potentially not inactivate all cells when using mitomycin C, as cells within clumps may not be exposed to the mitomycin C. Irradiated cells are preferred by those who have concerns about chemical treatment. Mitomycin C-treated cells are preferred by those who have concerns about DNA damage from irradiation.

What are the differences between CF1, CF6, and DR4 mouse embryonic fibroblasts?

CF1 mouse embryonic fibroblasts do not have drug resistance. CF6 mouse embryonic fibroblasts are resistant to Neomycin/geneticin (G418). DR 4 mouse embryonic fibroblasts are resistant to geneticin (G418), puromycin, hygromycin, and 6-thioguanine.

What is the recommended seeding density for mouse embryonic fibroblasts? Why is the seeding density critical?

We recommend seeding these cells at densities ranging from 2 x 10E4 to 5.5 x 10E4 cells/cm2. A good starting point is 3 x 10E4 cells/cm2. If the feeder cells are too sparse, they may not maintain the pluripotent cells without differentiation, and the pluripotent cells may not attach well. If the feeder cells are too dense, the feeder layer may detach from the plate, and the culture will be lost.

When should mouse embryonic fibroblasts be plated and how long can these cells be used?

These cells should be plated 24 hours prior to plating the ESCs or iPSCs and should be used for only 7-10 days.

Does the culture vessel need to be coated prior to seeding mouse embryonic fibroblasts?

Yes, the culture vessel needs to be coated with Attachment Factor protein (Cat. No. S006100) at 37 degrees C for 30 mins or at room temperature for 2 hours. The coated vessels can be used immediately or stored at room temperature for up to 24 hours.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.