Learn More
Gibco™ DPBS (10X), no calcium, no magnesium

Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).
We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Color | Clear |
| Concentration | 10 X |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 6.7 - 7.0 |
| Physical Form | Liquid |
| Product Line | Gibco™ |
| Quantity | 500 mL |
| Solution Type | Dulbecco's Phosphate Buffered Saline |
| Sterility | Sterile-filtered |
| Show More |
Safety and Handling
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.