Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ DPBS, calcium, magnesium
GREENER_CHOICE

Catalog No. p-4924946
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
10 x 500 mL
6 x 1000 mL
Osmolality:
270 to 300 mOsm/kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Osmolality
14-040-182 10 x 500 mL 270 to 300 mOsm/kg
14-040-141 100 mL 270 to 300 mOsm/kg
14-040-133 500 mL 270 to 300 mOsm/kg
14-040-117 1000 mL 270 to 300 mOsm/kg
14-040-216 6 x 1000 mL 270 to 300 mOsm/kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-040-182 Supplier Gibco™ Supplier No. 14040182
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

With: Calcium, Magnesium

Without: Phenol Red

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 1X
For Use With (Application) Washing and Staining Cells
pH 7.0 to 7.2
Physical Form Liquid
Product Line Gibco
Quantity 10 x 500 mL
Solution Type Cleaning, Storage, pH Buffer
Sterility Sterile
Includes No Sodium Pyruvate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Sterilization Method Sterile-filtered
Osmolality 270 to 300 mOsm/kg
Grade Cell Culture
Show More Show Less
Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.