Learn More
Gibco™ DPBS, no calcium, no magnesium
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 1X |
| For Use With (Application) | Mammalian Cell Culture |
| pH | 7.0 to 7.3 |
| Physical Form | Liquid |
| Product Line | Gibco |
| Quantity | 10 L |
| Solution Type | Cleaning, Storage, pH Buffer |
| Packaging | Bag |
| Sterility | Sterile |
| Show More |
Frequently Asked Questions (FAQs)
We cannot provide an official expiration date for open bottles as this depends on the customer's use. Once opened, Thermo Fisher Scientific can no longer guarantee the performance of any product. However, if the storage conditions are followed as recommended, the stated expiration date (36 months from the date of manufacture) should apply to opened bottles as DPBS is extremely stable.
DPBS has a shelf life of 36 months from the date of manufacture, when stored as recommended (15-30 degrees C).
The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.
We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.
Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.
Safety and Handling
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.