Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Geneticin™ Selective Antibiotic (G418 Sulfate) (50 mg/mL)

Catalog No. p-4918403
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-131-035 20 mL
10-131-027 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-131-035 Supplier Gibco™ Supplier No. 10131035
Only null left
Add to Cart
Edge
Add to Cart

Produced by bacterium Micromonospora rhodorangea and acts by binding ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells

Gibco Geneticin Selective Antibiotic (G418 Sulfate) is produced by the bacterium Micromonospora rhodorangea and acts by binding the ribosome, thus inhibiting protein synthesis in both prokaryotic and eukaryotic cells.

Resistance to Geneticin Selection Antibiotic (G418 Sulfate) is conferred by the E. coli APH(3)–I and APH(3)–II resistance genes.

  • Used as selective antibiotic in concentration range of 100 - 200μg/mL for bacteria, or 200 – 500μg/mL for most mammalian cells
  • Supplied as 50mg/mL solution in water

Cell Culture, Cloning, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Selection, Sf9 & Sf21 Cell Culture, Transfection, Transformation

Order Info

Shipping Condition: Room Temperature

Compliance

Manufactured at a cGMP compliant facility, which is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2 to 8°C
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture
Cell Type Eukaryotic Cells, Prokaryotic Cells
Concentration 50 mg/mL
Culture Type Mammalian Cell Culture, Insect Cell Culture
Form Liquid
Product Type Antibiotic
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
For Use With (Application) Eukaryotic Selection/Stable Cell Line Generation
Product Line Geneticin
Quantity 20 mL
Shipping Condition Room Temperature
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.