Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Grace's Insect Medium (2X), supplemented
GREENER_CHOICE

Catalog No. 11667037
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-667-037 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-667-037 Supplier Gibco™ Supplier No. 11667037
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in the agarose overlay when performing the plaque assay

Grace's Supplemented Insect Medium was originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. Used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS). Modified as follows: With Lactalbumin hydrolysate, Yeastolate, L-glutamine and Sodium bicarbonate. The complete formulation is available. Grace's Supplemented Insect Medium, 2x contains lactalbumin hydrolysate and yeastolate not commonly found in the unsupplemented version commonly diluted with sterile water, Fetal bovine Serum and agarose before use in the agarose overlay, as described in the product manual.

Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Room temperature

Compliance

Gibco Grace's Supplemented Insect Medium, 2x is manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Form Liquid
Product Type Grace's Insect Medium
With Additives Glutamine, Lactalbumin Hydrolysate, Yeastolate
Product Line Gibco
Quantity 100 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
What additional supplements are required in Grace's Insect Cell Culture Medium, Unsupplemented to make a "complete" formulation that will support insect cell growth?

Grace's Insect Cell Culture Medium, Unsupplemented requires addition of 3.33 g/L of lactalbumin hydrolysate and 3.33 g/L of yeastolate for the “base” growth medium. For the “complete” growth medium that will support insect cell growth, 10% fetal bovine serum must also be added. This “complete” growth medium is also referred to as TNM-FH.

What medium do you recommend to grow Sf9 or Sf21 cells for performing plaque assays?

We would recommend using our Grace's Supplemented Insect Medium (Cat. No. 11667037) or SF-900 Medium (1.3X) (Cat. No. 10967032). Grace's Supplmented Insect Media is a 2X formulation intended for use in the agarose overlay when performing the plaque assay. The media has been modified with:
- Lactalbumin hydrolysate
- Yeastolate
- L-glutamine
- Sodium bicarbonate

Please go here for the complete formulation (http://www.thermofisher.com/us/en/home/technical-resources/media-formulation.72.html). SF-900 Medium (1.3X) is a complete serum-free low protein insect cell culture medium for Sf9 and Sf21 cells, which can be used for 1% agarose overlays used for plaque assays.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.