Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Grace's Insect Medium, unsupplemented
GREENER_CHOICE

Catalog No. 11595030
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-595-030 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-595-030 Supplier Gibco™ Supplier No. 11595030
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Useful during transfection of insect cells or for protein labeling studies with methionine

Designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. One supplemented, widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS). Life Technologies offers a variety of Gibco Media options for a range of insect cell culture applications. The complete formulation is available.It does not contain lactalbumin hydrolysate or yeastolate, commonly found in the supplemented version. Gibco Grace's Unsupplemented Insect Medium is also made without methionine for protein labeling studies.

  • Manufactured at cGMP compliant facility located in Grand Island, New York
  • The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards
  • Requires supplementation with 10% (FBS), 50 mg/L methionine, 3.33 g/L lactalbumin hydrolysate and 3.33 g/L yeastolate before use as a growth medium

Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Form Liquid
Product Type Grace's Insect Medium
With Additives Glutamine, Sodium Bicarbonate
Without Additives No Lactalbumin Hydrolysate, No Yeastolate, No Methionine
Product Line Gibco
Quantity 500 mL
Shipping Condition Room Temperature
Inorganic Salts Calcium, Magnesium
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Show More Show Less
For how long can I store Grace's Insect Medium, unsupplemented at room temperature?

We ship this product under “ambient” conditions throughout the year. We have performed shipping studies that shown components to be stable for at least 7 days at ambient temperatures. The important factor is to ensure that the media is not exposed to light at the warmer temperatures as there are vitamins and amino acids that will breakdown with light exposure.

What additional supplements are required in Grace's Insect Cell Culture Medium, Unsupplemented to make a "complete" formulation that will support insect cell growth?

Grace's Insect Cell Culture Medium, Unsupplemented requires addition of 3.33 g/L of lactalbumin hydrolysate and 3.33 g/L of yeastolate for the “base” growth medium. For the “complete” growth medium that will support insect cell growth, 10% fetal bovine serum must also be added. This “complete” growth medium is also referred to as TNM-FH.

Do you offer a methionine-free media for baculovirus expression?

Yes, we offer our methionine-free Grace's Insect Medium (Cat. No. 11595030).

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.