Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Grace's Insect Medium, unsupplemented, powder

Catalog No. 11300027
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-300-027 10 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-300-027 Supplier Gibco™ Supplier No. 11300027
Get 15% Off Cell Culture Products
Use promo code CELLS15
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Requires supplementation with 10% Fetal Bovine Serum (FBS), 3.33g/L lactalbumin hydrolysate and 3.33g/L yeastolate before use as a growth medium as well as ambient CO2 in order to maintain physiological pH. Powder forms of Gibco cell culture medium require sodium bicarbonate supplementation, pH adjustment and filtration at the time of preparation.

Useful during transfection of insect cells or for protein labeling studies with methionine

Widely used for the growth of Spodeptera frugiperda cells, Sf9 and Sf21. Sf9 and Sf21 cells are routinely used to express recombinant protein using the Baculovirus Expression Vector System (BEVS). Was originally designed for the growth of cells from the Australian Emperor Gum Moth, Antherea eucalypti. It is modified as follows: with L-glutamine, without Lactalbumin hydrolysate, Yeastolate, Sodium bicarbonate. It does not contain lactalbumin hydrolysate and yeastolate commonly found in the supplemented version

Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture

Order Info

Shipping Condition: Room temperature

Compliance

Manufactured at a cGMP compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Insect Cell
Form Powder
Product Type Grace's Insect Medium
With Additives Glutamine
Without Additives No HEPES, No Phenol Red, No Sodium Pyruvate, No Lactalbumin Hydrolysate, No Yeastolate, No Sodium Bicarbonate
Product Line Gibco
Quantity 10 x 1 L
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
What additional supplements are required in Grace's Insect Cell Culture Medium, Unsupplemented to make a "complete" formulation that will support insect cell growth?

Grace's Insect Cell Culture Medium, Unsupplemented requires addition of 3.33 g/L of lactalbumin hydrolysate and 3.33 g/L of yeastolate for the “base” growth medium. For the “complete” growth medium that will support insect cell growth, 10% fetal bovine serum must also be added. This “complete” growth medium is also referred to as TNM-FH.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.