Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Leibovitz's L-15 Medium, powder

Catalog No. p-4919864
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 L
10 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-300-039 10 x 1 L
41-300-070 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-300-039 Supplier Gibco™ Supplier No. 41300039
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Requires

Supplementation

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue.

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.

This L-15 is modified as follows:

With: Galactose, Phenol Red, L-glutamine, Sodium Pyruvate

Without: Glucose, HEPES, Sodium Bicarbonate

The complete formulation is available.

L-15 contains no proteins, lipids, or growth factors. Therefore, F-10 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). Powder forms of Gibco™ L-15 cell culture medium require pH adjustment and filtration at the time of preparation (see protocol for details).

Please note that the shelf life on delivered products varies. This product has a minimum shelf life of 6 months at time of shipment. For more information regarding product shelf life, including details on specific product lots, please contact Customer Service.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Line HEP-2 monkey kidney cells
Cell Type Explants of embryonic and adult human tissue
Classification Animal Origin
Form Powder
Product Type Leibovitz's L-15 Medium
Sterility Sterile-filtered
With Additives Glutamine, Phenol Red, Sodium Pyruvate, Galactose
Without Additives No Glucose, No HEPES, No Sodium Bicarbonate
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
Product Line Gibco
Quantity 10 x 1 L
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.