Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Liver Cancer Cell Culture Starter Kit, North America

Catalog No. A3518201
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3518201 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3518201 Supplier Gibco™ Supplier No. A3518201
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gibco Cancer Cell Culture Starter kits provide researchers with a convenient package of reagents for the culture and transfection of cancer cell types frequently used in cancer research. Cancer Cell Culture Starter kits are currently available for breast, lung, prostate, and liver cancer cells.

Each starter kit includes the following products for cancer cell culture and transfection:

  • Basal media
  • Fetal bovine serum
  • Penicillin-streptomycin
  • TrypLE Express Enzyme
  • Lipofectamine 3000 Transfection Reagent
  • Opti-MEM I
  • Nunc Cell Culture Treated EasYFlasks

The Cancer Cell Culture Starter kits contain the most commonly used reagents to help you start your cancer cell culture quickly and achieve successful transfections, based on a review of hundreds of cancer research publications in which the following cell lines were used:

  • Prostate cancer: DU145, PC-3, LNCaP
  • Lung cancer: A549, NCI-H975, PC-14, NCI-H1299
  • Breast cancer: MCF-7, MDA-MB-231, T-47D, BT-474
  • Liver cancer: Hep G2, Hep 3B

The Cancer Culture Starter kits also include a suggested protocol for the culture, dissociation (cell passage), and transfection of the most-published cancer cell lines. Achieve peace of mind and quickly initiate your experiments with these starter kits, the components of which may also be purchased separately.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cell Culture Starter Kit
Quantity 1 Kit
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Content And Storage • DMEM, high glucose, with GlutaMAX Supplement (store at 2–8°C)
• Fetal bovine serum, qualified, One Shot™ format, US origin (store at -5 to -30°C)
• Penicillin-streptomycin (10,000 U/mL) (store at -5 to -30°C)
• TrypLE Express Enzyme (1 X), no phenol red (store at room temperature)
• Lipofectamine 3000 Transfection Reagent (store at 2–8°C)
• Opti-MEM I Reduced Serum Medium (store at 2–8°C)
• 1 case of 100 Nunc Cell Culture Treated EasYFlasks

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.