Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ PFHM-II Protein-Free Hybridoma Medium (liquid)
GREENER_CHOICE

Catalog No. 12040077
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-040-077 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-040-077 Supplier Gibco™ Supplier No. 12040077
  • $142.00
    $131.65 / Each of 1

    Save $10.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Serum-free, protein-free medium specifically developed for the ability to support the growth of hybridomas for antibody production

  • Ability to support suspension and stationary hybridomas and myelomas
  • Complete, ready-to-use, serum-free, protein-free formulation
  • Ability to support suspension and stationary hybridomas and myelomas
  • Suitable for culture of recombinant myeloma lines as well as traditional hybridomas, Gibco PFHM II provides high cell densities and IgG production
  • Cholesterol-dependent cultures such as NS0 require the addition of 250x Cholesterol Lipid Concentrate
  • Sustainable packaging

Antibody and Cell Culture Production, Bioproduction, Cell Culture, Mammalian Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hybridomas
Classification Protein-free
Form Liquid
Product Type Hybridoma Medium
Serum Level Serum-free
With Additives Glutamine, Phenol Red
Product Line Gibco
Quantity 1000 mL
Shipping Condition Room Temperature
Content And Storage Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Culture Type Suspension Cell Culture
Show More Show Less
How do I adapt my cells to serum-free medium?

Cells can be adapted by Sequential or Direct Adaptation. Suggested protocols for each are below, and you can also find more information by searching "Adaptation of Cell Cultures to a Serum-Free Medium" from our website home page.

SEQUENTIAL ADAPTATION
1) Subculture the cells growing in serum-supplemented medium into a 25%:75% mixture of SFM and serum supplemented medium.
2) When the cell density is 5 x 10E5 cells/ml, subculture the cells into a 50%:50% mixture of SFM and serum supplemented medium at a cell density 2.5 x 10E5 to 3 x 10E5 cells/ml.
3) Continue to subculture after the cell density 5 x 10E5 cells/ml in gradually increasing proportions of SFM until the serum is ~0.1% with about 85% cell viability.
4) Subculture the cells into SFM with an innoculum of 2.5 x 10E5 to 3 x 10E5 cells/ml.
5) When the cell density is 1 x 10E6 to 3 x 10E6 cells/ml (4 to 6 days post planting) subculture the cells again.
6) Stock cultures of SFM adapted cells should be subcultured in SFM every 3 to 5 days when the cell density is 1 x 10E6 to 3 x 10E6 cells/ml with 90% viability.

DIRECT ADAPTATION
Some cells can be directly adapted from serum-containing medium to SFM. For direct adaptation, the cell innoculum should be 1.5 x 10E5 to 3 x 10E5 cells/ml.
Cells should be subcultured when the cell density is 1 x 10E6 to 3 x 10E6 cells/ml. Cells are fully adapted to SFM when the cell density is 2 x 10E6 to 4 x 10E6 cells/ml after 4 to 7 days in culture.
Stock cultures of cells adapted to SFM should be subcultured in SFM every 3 to 5 days when the cell density is 1 x 10E6 to 3 x 10E6 cells/ml with 90% viability.

Why is it necessary to gradually adapt the cells to serum-free medium?

Some cells, such as insect cells, are sensitive to changes in their medium. By sequentially adapting cells, the medium is changed with minimal effects on cell growth.

Can I freeze PFHM-II Protein-Free Hybridoma Medium (liquid) (Cat. No. 12040077)?

We do not recommend freezing our liquid media products. Freezing can cause some components like amino acids and inorganic salts to precipitate upon thawing.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.