Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Sf-900™ III SFM

Catalog No. 50591352
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1000 mL
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-591-352 20 L
12-658-019 500 mL
12-658-027 1000 mL
12-658-035 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-591-352 Supplier Gibco™ Supplier No. 12658001
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium

Sf-900 III SFM is a low-hydrolysate, serum-free, protein-free, animal origin–free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells.

  • Optimized for recombinant gene expression using the baculovirus and stable insect expression systems
  • Suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines
  • Spodoptera frugiperda (Sf9) cells grown in Sf-900 III SFM achieve maximum cell densities of 10–14 x 106 cells/mL
  • An improved serum-free, protein-free, animal origin–free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect viruses and recombinant proteins
  • Ready to use format does not require addition of serum, glutamine, or surfactants
  • Cells adapted to other commercially available serum-free media should be sequentially adapted into Sf-900 III SFM
  • Reduced hydrolysate concentrations improve lot-to-lot consistency

Order Info

Shipping Condition: Room temperature

Compliance

Gibco Sf-900 II SFM is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Sf21, Sf9
Cell Type Insect Cell
Classification Animal Origin-free
Concentration 1 X
Form Liquid
Product Type Insect Cell Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile
With Additives Glutamine
Without Additives No Phenol Red, No Sodium Pyruvate, No Sodium Bicarbonate
Product Line Gibco, Sf-900
Quantity 20 L
Shipping Condition Room Temperature
Endotoxin Level Low
Purity or Quality Grade Cell Culture Grade
Content And Storage Storage conditions: 2–8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 18 months from date of manufacture
Culture Environment Non-CO2
Culture Type Suspension Cell Culture
Show More Show Less
What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.