Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Sf21 cells in Sf-900™ III SFM

Catalog No. 12682019
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-682-019 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Review the Material Safety Data Sheet before handling.
Catalog No. 12-682-019 Supplier Gibco™ Supplier No. 12682019
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Review the Material Safety Data Sheet before handling.

Used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins

Gibco Sf21 cells are adapted to serum-free suspension culture in Gibco Sf-900™ III SFM, which saves significant time and expense associated with the adaptation of cultures.

  • Recombinant protein expression from a variety of expression systems
  • Good growth in adherent or suspension culture
  • Documented lineage from a low passage Master Cell Bank
  • Quality and performance testing

Baculovirus Production and Titering, Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: Liquid nitrogen (vapor phase)
Shipping conditions: Frozen
Cell Line Sf21
Cell Type Insect Cells
Species S. frugiperda
Media Recommendation Sf-900 III SFM (Serum-Free Media)
Product Line Sf-900
Product Type Insect Cells
Quantity 1 Vial
Shipping Condition Dry Ice
What is the procedure to thaw frozen insect cells?

The following protocol describes a general procedure for thawing cryopreserved cells. For detailed protocols, always refer to the cell-specific product insert.

1. Remove the cryovial containing the frozen cells from liquid nitrogen storage and immediately place it into a 37°C water bath.
2. Quickly thaw the cells (< 1 minute) by gently swirling the vial in the 37°C water bath until there is just a small bit of ice left in the vial.
3. Transfer the vial into a laminar flow hood. Before opening, wipe the outside of the vial with 70% ethanol.
4. Transfer the desired amount of pre-warmed complete growth medium appropriate for your cell line dropwise into the centrifuge tube containing the thawed cells.
5. Centrifuge the cell suspension at approximately 200 x g for 5-10 minutes. The actual centrifugation speed and duration varies depending on the cell type.
6. After the centrifugation, check the clarity of supernatant and visibility of a complete pellet. Aseptically decant the supernatant without disturbing the cell pellet.
7. Gently resuspend the cells in complete growth medium, and transfer them into the appropriate culture vessel and into the recommended culture environment.

Note: The appropriate flask size depends on the number of cells frozen in the cryovial, and the culture environment varies based on the cell and media type.

Why does the Insect cell line manual state: "Cells should be maintained at 27 degrees C in a non-humidified environment."

Insect cells do not require CO2 or high humidity to grow, they can grow in a lab drawer at room temperature. We recommend this so people don't waste CO2 and other resources necessary for maintaining a tissue culture incubator. It should be noted, however, that the cells will grow in a humidified incubator.

What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

What methods can be used to detach tightly adherent Sf9 and Sf21 cells cultured in Grace's media supplemented with 10% heat-inactivated FBS, Sf-900 II SFM, or Sf-900 III SFM?

Sf9 and Sf21 cells should be lightly adherent cells. However, there are some Sf9 and Sf21 cells that attach to culture vessels very tightly. The use of enzymes such as trypsin, collagenase, hyaluronidase, TrypLE Express, and TrypLE Select have been tried without success for passaging cells. The main problem is that the cells do not attach well after having dissociated with the enzymes.

The best method to use is to culture cells in a T-flask. Close cap tightly and hold flask with cap pointing towards the ceiling. Hit the bottom of the flask over a counter 2-3 times with medium force. Cell detachment may be 60-80% and not 100%. This will allow for detachment of enough cells for passaging. If tapping the flask over the counter is performed with too harsh of a force or too many times, cell viability will be greatly affected.

If possible, we recommend that you culture cells in suspension conditions. Cells in suspension cultures can be passaged directly into adherent conditions when needed. The culture of cells in suspension conditions will allow for higher cell densities as cell growth is not limited to the surface area.

How can I concentrate my insect cells to increase the cell density?

If the cell density is too low and the cells have been in culture for 4-5 days, we recommend concentrating the cells by centrifuging them at 100 X g for 5 minutes and resuspending them in fresh medium. Cells should not be left in the same medium for more than 4-5 days as nutrients in the medium will have been used up by the cells in that period, and the medium itself degraded due to prolonged exposure to warm temperatures. Cells should also be centrifuged and concentrated if a lot of cell debris is observed in culture.

What are the main differences between insect cell culture and mammalian cell culture?

Insect cells are much more fragile than a lot of mammalian cell lines. They suffer much more damage than mammalian cells from overgrowth and over-splitting. Never let cells go above 8 x 10E6 cells/mL or grow at densities less than 0.5 x 10E6 cells/mL in suspension. Insect cells require a little more osmotic pressure than mammalian cells (340 µOsM). Insect cells use a lot of O2, especially during protein expression. Insect cell culture media is more acidic than mammalian media (pH 6.0-6.4). The insect cell culture media is phosphate buffer based. Therefore, no CO2 is needed to maintain the pH.

Which insect cell line is best to use?

We recommend Sf9 or Sf21 cells for transfection, purification, and amplification of recombinant virus. Sf9 cells are regular in size, easy to manipulate, and form good monolayers for plaque assays. Sf9 and Sf21 cells can also be used for expression of recombinant proteins, but the High Five cell line may produce higher levels. We recommend the High Five cell line for expression of secreted recombinant proteins. They are grown in serum-free medium, adaptable to suspension culture, and produce high levels of recombinant protein (Davis et al., 1992; see http://www.ncbi.nlm.nih.gov/pubmed/1368794).

Note: Generally it is easier to use one cell line for procedures up to optimization of protein expression. Once you have confirmed expression of your recombinant protein, other cell lines can be tried for optimization of expression levels.

Can I grow insect cells for short periods at 37 degrees C?

No, this is not recommended. Prolonged exposure to temperature higher than 29 degrees C will cause cell death. It is better to grow the cells at 27 degrees C or room temperature.

Is CO2 required for insect cell culture?

No, CO2 exchange is not required for insect cell culture.

What is the best way to plate insect cells?

Prior to performing transfections and plaque assays, cells need to be evenly distributed over the surface of a tissue culture plate. This ensures that:
- Cells do not distribute unevenly, leading to asymmetric monolayers.
- Maximum cell surface area is available for infection.

To disperse cells:
1. Rock the flask or plate slowly by hand forward and backward, then side-to-side.
2. Repeat this four times, watching carefully to ensure that the liquid reaches all areas of the growth surface.
3. Do not use a circular motion to disperse the cells, because this causes a concentration of cells around the edges of the plate rather than an even distribution.

What are the doubling times for robust growth and maintenance of different insect cell lines?

Please see below for information related to the type of cells, media, and doubling time in hours:

D.mel-2; Schneider's Drosophila + 10% FBS heat-inactivated; 18 to 24
High Five cells; Express Five SFM; approximately 24
Sf9 cells; Sf-900 II SFM; 24 to 30
Sf9 cells; Sf-900 III SFM; 24 to 30
Sf21 cells; Sf-900 II SFM; 24 to 30
Sf21 cells Sf-900 III SFM; 24 to 30

What are the sizes of Sf9, Sf21, High Five cells, and D.mel-2 cells?

Please see below for information related to type of cells, media, and cell size in microns:

D.mel-2; Schneider's Drosophila + 10% FBS heat-inactivated; 10 to 12
High Five cells; Express Five SFM; 17.5 to 19.5
Sf9 cell; Sf-900 II SFM; 15 to 17.5
Sf9 cells; Sf-900 III SFM; 15 to 17.5
Sf21 cell;s Sf-900 II SFM; 15 to 17.5
Sf21 cells; Sf-900 III SFM; 15 to 17.5


WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.