Learn More
TARGETMOL CHEMICALS INC GIBH-130 5mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T44505MG
Encompass_Preferred
GIBH-130 markedly inhibits the IL-1β secretion by activated microglia (IC50: 3.4 nM). GIBH-130 is an effective inhibitor of neuroinflammation.
Catalog No. NC1624930