Learn More
New England Biolabs, Inc. Gibson Assembly Cloning Kit – 10 reactions
SDP
Catalog No. NC0414992
Encompass
Gibson Assembly allows for successful assembly of multiple DNA fragments, regardless of fragment length or end compatibility. The cloning kit contains NEB 5-alpha Competent Cells.
Catalog No. NC0414992 Supplier New England Biolabs, Inc. Supplier No. E5510S
Add to Cart
Add to Cart
Gibson Assembly allows for successful assembly of multiple DNA fragments, regardless of fragment length or end compatibility. The cloning kit contains NEB 5-alpha Competent Cells.