Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibson Bioscience Reagents: Catalese

Catalog No. 21200533
Change view
Click to view available options
Product Type:
Catalase Reagent
Kit Size:
2.5mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Kit Size
21-200-533 Catalase Reagent 50mL
21-200-326 Catalase Reagent 2.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-200-533 Supplier Gibson Laboratories LLC Supplier No. 60004LEX

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the detection of microorganisms that possess the catalase enzyme

Specifications

Kit Size 50mL
Product Type Catalase Reagent
Formulation 1L hydrogen peroxide (3%)
For Use With (Application) Detection of microorganisms that possess the catalase enzyme
Storage Requirements 2° to 25°C
Volume (Metric) 50mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.