Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibson Bioscience Reagents: KOH 10%

Catalog No. 21200331
Change view
Click to view available options
Kit Size:
10 × 2.5mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Kit Size
21-200-331 10 × 2.5mL
21-200-315 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-200-331 Supplier Gibson Laboratories LLC Supplier No. 300145LEX

missing translation for 'validMainframeMsg'

For indicated test(s)

Specifications

Kit Size 10 × 2.5mL
Product Type KOH
Formulation Potassium hydroxide 100g, deionized water 1L
For Use With (Application) KOH prep for fungal elements
Storage Requirements 2° to 25°C
Volume (Metric) 2.5mL
Test Time 10 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.