Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibson Bioscience Reagents: Oxidase Test

Catalog No. 21200303
Change view
Click to view available options
Product Type:
Oxidase Test Swab
Kit Size:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Kit Size
21-200-303 Oxidase Test Swab 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-200-303 Supplier Gibson Laboratories LLC Supplier No. 60500LEX

missing translation for 'validMainframeMsg'

For the detection of oxidase producing organisms.

  • Features a rapid colorimetric test
  • Used for presumptive identification of cytochrome oxidase activity in microorganisms (Neisseria)
  • Helps in identification of Pseudomonas and other cytochrome oxidase-positive gram negative microorganisms

Specifications

Kit Size 50 Tests
Product Type Oxidase Test Swab
Formulation N-N-N-tetramethyl-p-phenylenediamine 10g, ascorbic acid 1g, deionized water 1L
For Use With (Application) Rapid colorimetric test for presumptive identification of cytochrome oxidase activity in microorganisms
Storage Requirements 2° to 25°C
Test Time 10 sec.
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.