Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibson Bioscience Reagents: PYR

Catalog No. 21200322
Change view
Click to view available options
Volume (Metric):
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-200-322 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-322 Supplier Gibson Laboratories LLC Supplier No. 60029LEX
This item is not returnable. View return policy

For use with PYR and LAP disks

Specifications

Product Type PYR Reagent
Formulation p-Dimethlyaminocinnamaldehyde 1.7g, methanol 0.3L, hydrochloric acid 0.3L, phosphate buffer 0.4L, pH 6.8
For Use With (Application) Use as an indicator reagent for the detection of pyrrolidonylarylamidase, an enzyme produced by Streptococcus pyogenes and Enterococcus faecalis
Storage Requirements Room Temperature
Volume (Metric) 50 mL
Test Time 3 to 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.