Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Veritor™ Plus Analyzer with Flu A+B CLIA Waived Flu Assay Kits (2 x 30)

Catalog No. b256074 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
2 x 30 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B256074 2 x 30 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Dispose of used BD Veritor System test devices as biohazardous waste.
Catalog No. B256074 Supplier BD Supplier No. 256074
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dispose of used BD Veritor System test devices as biohazardous waste.

Point of Care lab-quality detection of influenza A and B. BD Veritor™ Plus Analyzer with Flu A+B CLIA Waived Flu Assay Kits (2 x 30) includes an advanced chromatographic instrument and two immunoassay kits to detect antigens from respiratory specimens of symptomatic patients in 10 minutes.

Veritor™ Plus Analyzer
This chromatographic instrument features advanced particle technology that enhances sensitivity and adaptive read technology to compensate for background and non-specific binding to reduce false-positive results. With Analyze Now and Walk-Away modes, the instrument provides lab-quality test results in just 10 minutes so you can make decisions while the patient is onsite.

  • Cleared for use with nasal swabs and nasopharyngeal swabs
  • Reduce hands on time and create a flexible workflow
  • Improved traceability with InfoScan module
  • Better connectivity with InfoSync module
  • Number of reads per instrument: 3500
  • Rechargeable lithium battery, print-enabled

Influenza A+B CLIA Waived Assay Kits
This system detects nucleoprotein antigens from nasal swab and nasopharyngeal swab in transport media samples from patients.

  • Aids in rapid diagnosis of influenza A and B viral infections
  • Tests samples processed from respiratory specimens
  • Sensitive and specific for test to reduce false positives
  • Components: Lateral flow test strips, RV reagent C in tubes (100μL per tube), 300μL pipettes, Flu A+/B- control swab, Flu B+/A- control swab
  • Two kits included, sufficient for 60 assays (2 x 30)
TRUSTED_SUSTAINABILITY

Specifications

Description Flu A + B Test Kit
Clia Complexity Waived
Detectable Analytes Influenza A and B
Sample Type Nasal swabs and nasopharyngeal (NP) swabs
Type Analyzer and Flu A+B Immunoassay Test Kit
For Use With (Application) Detection of Flu A + B
Includes Analyzer with lithium battery, two assay kits including: Lateral flow test strips, Pipettes, Flu A+/B- control swab, Flu B+/A- control swab, RV reagent C in tubes
Packaging Type 1 Kit
Quantity 2 x 30 Assays
Test Time 10 min.

For in vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.