Learn More
Diagnostic Biosystems GIEMSA Giemsa
Catalog No. 502407568
Encompass_Preferred
The Giemsa Stain Kit (May-Grunwald) is intended for use in the visualization of cells present in hematopoietic tissues and certain microorganisms. This kit may be used on formalin-fixed, paraffin-embedded sections.
Catalog No. 50-240-7568 Supplier Diagnostic Biosystems Supplier No. KT016
The Giemsa Stain Kit (May-Grunwald) is intended for use in the visualization of cells present in hematopoietic tissues and certain microorganisms. This kit may be used on formalin-fixed, paraffin-embedded sections.