Learn More
Newcomer Supply Giemsa Control Slides, 10/set
SDP
Catalog No. NC0915321
Encompass
Newcomer Supply Giemsa Control Slides are for the positive histochemical and differential staining of hematopoietic tissue. Giemsa Control Slides contain sections of positive staining spleen.
Catalog No. NC0915321 Supplier Newcomer Supply Supplier No. 4240A
Newcomer Supply Giemsa Control Slides are for the positive histochemical and differential staining of hematopoietic tissue. Giemsa Control Slides contain sections of positive staining spleen.