Learn More
Epredia™ Giemsa Stain
Catalog No. 22050290 Shop All Epredia Products
Change view
Click to view available options
Quantity:
32 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-050-290 1 gal.
22-050-291 32 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-050-290 Supplier Epredia™ Supplier No. 89004
Add to Cart
Add to Cart
This item is not returnable. View return policy
Take advantage of an excellent classical blood film stain used on peripheral blood smears and bone marrow with the Epredia™ Giemsa Stain.
  • Erythrocytes stain pink with platelets exhibiting a very light, pale hue
  • Leukocyte nuclear chromatin stains magenta-purple
  • Monocyte cytoplasm stains the classical “ground glass” blue
  • Lymphocyte cytoplasm stains clear, sky blue
Specifications
Quantity 1 gal.
Product Type Giemsa Stain
WARNING: Reproductive Harm - www.P65Warnings.ca.gov