Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giemsa Stain (Crystalline), Fisher BioReagents

Catalog No. BP24225 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP24225 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP24225

missing translation for 'validMainframeMsg'

Giemsa is a biological stain used for thin blood films to differentiate leukocytes, for staining malarial parasites in thin and thick blood films, and for staining bone marrow to show cell morphology.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51811-82-6
Molecular Formula C14H14ClN3S
Molecular Weight (g/mol) 291.80
MDL Number MFCD00012112,MFCD00081642
InChI Key NALREUIWICQLPS-UHFFFAOYSA-N
PubChem CID 13735
IUPAC Name (7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1

Specifications

Melting Point 300°C
Bio. Stain Comm. Certification Pass Test
Identification Pass Test
Formula Weight 291.8
Packaging Glass Bottle
Color Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Giemsa Stain
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.