Learn More
Scytek Laboratories Giemsa Stain Kit (May-Grunwald): Large
SDP
Catalog No. NC1969848
Encompass_Preferred
The Giemsa Stain Kit (May-Grunwald) is intended for use in the visualization of cells present in hematopoietic tissues and certain microorganisms. This kit may be used on formalin-fixed, paraffin-embedded sections.Nuclei: Blue/VioletCytoplasm: Light BlueCollagen: Pale PinkMuscle Fibers: Pale PinkErythrocytes: Gray, Yellow or PinkRickettsia: Reddish-PurpleHelicobacter Pylori: BlueMast Cells: Dark Blue with Red GranulesKit contents: MAY500, May-Grunwald Stock Solution, 500 ml. GGS500, Giemsa Stock Solution, 500 ml. PBM500, Phosphate Buffer Solution, pH 6.8, 500 ml.
This item is not returnable. View return policy
Catalog No. NC1969848 Supplier Scytek Laboratories Supplier No. GMG1
The Giemsa Stain Kit (May-Grunwald) is intended for use in the visualization of cells present in hematopoietic tissues and certain microorganisms. This kit may be used on formalin-fixed, paraffin-embedded sections.Nuclei: Blue/VioletCytoplasm: Light BlueCollagen: Pale PinkMuscle Fibers: Pale PinkErythrocytes: Gray, Yellow or PinkRickettsia: Reddish-PurpleHelicobacter Pylori: BlueMast Cells: Dark Blue with Red GranulesKit contents: MAY500, May-Grunwald Stock Solution, 500 ml. GGS500, Giemsa Stock Solution, 500 ml. PBM500, Phosphate Buffer Solution, pH 6.8, 500 ml.