Learn More
Scytek Laboratories Giemsa Stain Kit (May-Grunwald): Small
SDP
Catalog No. NC1969849
Encompass_Preferred
The Giemsa Stain Kit (May-Grunwald) is intended for use in the visualization of cells present in hematopoietic tissues and certain microorganisms. This kit may be used on formalin-fixed, paraffin-embedded sections.Nuclei: Blue/VioletCytoplasm: Light BlueCollagen: Pale PinkMuscle Fibers: Pale PinkErythrocytes: Gray, Yellow or PinkRickettsia: Reddish-PurpleHelicobacter Pylori: BlueMast Cells: Dark Blue with Red GranulesKit contents: MAY030, May-Grunwald Stock Solution, 30 ml. GGS008, Giemsa Stock Solution, 8 ml. PBM060, Phosphate Buffer Solution, pH 6.8, 2 x 60 ml.
This item is not returnable. View return policy
Catalog No. NC1969849 Supplier Scytek Laboratories Supplier No. GMG2
The Giemsa Stain Kit (May-Grunwald) is intended for use in the visualization of cells present in hematopoietic tissues and certain microorganisms. This kit may be used on formalin-fixed, paraffin-embedded sections.Nuclei: Blue/VioletCytoplasm: Light BlueCollagen: Pale PinkMuscle Fibers: Pale PinkErythrocytes: Gray, Yellow or PinkRickettsia: Reddish-PurpleHelicobacter Pylori: BlueMast Cells: Dark Blue with Red GranulesKit contents: MAY030, May-Grunwald Stock Solution, 30 ml. GGS008, Giemsa Stock Solution, 8 ml. PBM060, Phosphate Buffer Solution, pH 6.8, 2 x 60 ml.