Learn More
StatLab Giemsa Stain Kit - Optimized
Description
- Pint stain kit yields between 340-620 slides
- The MasterTech Optimized Giemsa Stain Kit restores these metachromatic qualities within the dye through the development of a specially formulated Giemsa Buffer, providing exceptional staining results
- Giemsa serves as the primary stain, producing blue to purple nuclei, cytoplasm, and helicobacter
- Pint Kit Consists of: Giemsa Stain Stock Pint and Optimized Giemsa Buffer 5 Pints
Specifications
Specifications
| Product Type | Stain Kit |