Learn More
Giemsa Stain, Modified Solution, Sigma-Aldrich
Catalog No. 01259022 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-022 1 L
01-258-846 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-022 Supplier MilliporeSigma Supelco Supplier No. 489001LF
Add to Cart
Add to Cart
For research use only, for the staining of cellular blood components and blood parasites.
When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration. The use of purified eosin and thiazine dyes minimizes lot-to-lot variation.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 L
Product Type Giemsa Stain