Learn More
Electron Microscopy Sciences Giemsa Stain Powder, Certified C.I. DcGe-19 10 GR
SDP
Catalog No. 50980522
Encompass_Preferred
Thin film stain for differentiation of types of leucocytes; Rickettsiae, bacteria, & inclusion bodies. May-Grunwald/Giemsa for bone marrow stain. Pinkus' acid orcein-giemsa for connective tissue staining. Certified by the Biological Stain Commission All Dyes/Stains are for research use only. Not meant for drug, food household use.
Catalog No. 50-980-522 Supplier Electron Microscopy Sciences Supplier No. 15942
Thin film stain for differentiation of types of leucocytes; Rickettsiae, bacteria, & inclusion bodies. May-Grunwald/Giemsa for bone marrow stain. Pinkus' acid orcein-giemsa for connective tissue staining. Certified by the Biological Stain Commission All Dyes/Stains are for research use only. Not meant for drug, food household use.