Learn More
Apexbio Technology LLC Giemsa Staining Solution (10X) 500ml
SDP
Catalog No. 5000063341
Encompass_Preferred
Giemsa staining solution a mixture primarily composed of Azure dyes and Eosin is widely utilized in biological research for cellular identification and differentiation based on chemical interactions between dye components and cellular structures Azure dyes preferentially bind to nuclear elements and acidic cytoplasmic components (e g RNA) whereas Eosin selectively stains alkaline cytoplasmic components such as hemoglobin granules Typically applied at a 1X final concentration after dilution this method facilitates distinct visualization of blood smear samples peripheral blood leukocyte differentiation identification of parasites as well as chromosome banding after appropriate pretreatment Cytoplasmic structures stained with Giemsa appear pink to bluish nuclei stain purple or bluish-purple enabling clear differentiation of erythrocytes various leukocyte subsets and chromosome bands in cytogenetic analyses
Catalog No. 50-000-63341 Supplier Apexbio Technology LLC Supplier No. K1181500
Giemsa staining solution a mixture primarily composed of Azure dyes and Eosin is widely utilized in biological research for cellular identification and differentiation based on chemical interactions between dye components and cellular structures Azure dyes preferentially bind to nuclear elements and acidic cytoplasmic components (e g RNA) whereas Eosin selectively stains alkaline cytoplasmic components such as hemoglobin granules Typically applied at a 1X final concentration after dilution this method facilitates distinct visualization of blood smear samples peripheral blood leukocyte differentiation identification of parasites as well as chromosome banding after appropriate pretreatment Cytoplasmic structures stained with Giemsa appear pink to bluish nuclei stain purple or bluish-purple enabling clear differentiation of erythrocytes various leukocyte subsets and chromosome bands in cytogenetic analyses