Learn More
Electron Microscopy Sciences Giemsa Stain Solution C.I. DcGe-19 100 ML
SDP
Catalog No. 50980520
Encompass_Preferred
References: Differentiates types of leucocytes, bacteria and inclusion bodies. Giemsa, Centralbl. F. Publ., Abt. 1,37, 308 (1904) Stain for thick blood films; shows malarial parasites. Barber and Komp, Publ. Health Rep., 44, 2330 (1929) Bone marrow and stain for showing cell morphology; together with May-Grunwald Solution for pap staining. Pappenheim, Folia Haemat., 13, 338 (1912).
Catalog No. 50-980-520 Supplier Electron Microscopy Sciences Supplier No. 15940
References: Differentiates types of leucocytes, bacteria and inclusion bodies. Giemsa, Centralbl. F. Publ., Abt. 1,37, 308 (1904) Stain for thick blood films; shows malarial parasites. Barber and Komp, Publ. Health Rep., 44, 2330 (1929) Bone marrow and stain for showing cell morphology; together with May-Grunwald Solution for pap staining. Pappenheim, Folia Haemat., 13, 338 (1912).