Learn More
Giemsa Stain, Technical Grade, Sigma-Aldrich
Catalog No. 01258931 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
01-258-931 25 g
1 options
Catalog No. 01-258-931 Supplier MilliporeSigma Supelco Supplier No. G450725G
Add to Cart
Add to Cart
Differential stain used for blood and bone marrow specimens, clinical cytology, bacteriology, histological biopsies, and tumor samples.
Giemsa stain is a versatile differential stain that is widely used in hematology for blood and bone marrow specimens, clinical cytology specimens, bacteriology, histological biopsies, and tumor samples. Giemsa's staining solution is composed of methylene blue, azure, and eosin. It is a major histological stain due to its unique staining of chromatin, nuclear membranes, and cytoplasmic elements. The stain is frequently used in combination with other dye solutions: May-Grunwald's solution for Pappenheim (MGG) and Wright-Giemsa.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Product Type Giemsa Stain